Learn More
Aviva Systems Biology Corp SMG5 Antibody - N-terminal region (ARP55229_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55229_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMG5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMG5. Alternate names: EST1B, FLJ34864, KIAA1089, LPTS-RP1, LPTSRP1, RP11-54H19.7, SMG-5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-61441