Learn More
Lonza Walkersville SmGM- 2 Smooth Muscle Cell Growth Medium -2 BulletKit
Catalog No. NC9460389
Encompass
SmGM-2 Bulletkit is also sold as separate basal medium and supplements.SmGM-2 Smooth Muscle Growth Medium-2 is a robust medium that was developed for the maintenance and expansion of human smooth muscle cells from various tissue origins including cardiac, airway, prostate and more. This kit is also utilized to support the maintenance of intestinal cells. SmGM-2 medium contains 5% FBS.Clonetics Media Products are specially designed to support the growth of human and animal primary derived cells. Media systems have been specifically developed and optimized for various cell systems.Content:1 x SmBM Basal Medium (CC-3181), 500 mL1 x SmGM -2 SingleQuotsSupplement Pack (CC-4149) containing:1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Gray Cap Vial with hFGF-B, 1.00 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Bottle FBS, 25.00 mL1 x Green Cap Vial with hEGF, 0.50 mL
Catalog No. NC9460389 Supplier Lonza Walkersville Supplier No. CC3182
SmGM-2 Bulletkit is also sold as separate basal medium and supplements.SmGM-2 Smooth Muscle Growth Medium-2 is a robust medium that was developed for the maintenance and expansion of human smooth muscle cells from various tissue origins including cardiac, airway, prostate and more. This kit is also utilized to support the maintenance of intestinal cells. SmGM-2 medium contains 5% FBS.Clonetics Media Products are specially designed to support the growth of human and animal primary derived cells. Media systems have been specifically developed and optimized for various cell systems.Content:1 x SmBM Basal Medium (CC-3181), 500 mL1 x SmGM -2 SingleQuotsSupplement Pack (CC-4149) containing:1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Gray Cap Vial with hFGF-B, 1.00 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Bottle FBS, 25.00 mL1 x Green Cap Vial with hEGF, 0.50 mL