Learn More
Lonza Walkersville SmGM- 2 Smooth Muscle Cell Growth Medium-2 SingleQuots Supplements and Growth Factors
Catalog No. NC9650299
Encompass
SmGM -2 SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the SmGM -2 Smooth Muscle Cell Growth Medium BulletKit (Catalog Number CC-3182). The full SmGM -2 BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3182.SmGM-2 Smooth Muscle Growth Medium-2 is a robust medium that was developed for the maintenance and expansion of human smooth muscle cells from various tissue origins including cardiac, airway, prostate and more. This kit is also utilized to support the maintenance of intestinal cells. SmGM-2 medium contains 5% FBS.Content:1 x Lilac Cap Vial with Insulin 0.50 mL1 x Gray Cap Vial with hFGF-B 1.00 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Bottle FBS 25.00 mL1 x Green Cap Vial with hEGF 0.50 mL
Catalog No. NC9650299 Supplier Lonza Walkersville Supplier No. CC4149
SmGM -2 SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the SmGM -2 Smooth Muscle Cell Growth Medium BulletKit (Catalog Number CC-3182). The full SmGM -2 BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3182.SmGM-2 Smooth Muscle Growth Medium-2 is a robust medium that was developed for the maintenance and expansion of human smooth muscle cells from various tissue origins including cardiac, airway, prostate and more. This kit is also utilized to support the maintenance of intestinal cells. SmGM-2 medium contains 5% FBS.Content:1 x Lilac Cap Vial with Insulin 0.50 mL1 x Gray Cap Vial with hFGF-B 1.00 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Bottle FBS 25.00 mL1 x Green Cap Vial with hEGF 0.50 mL