Learn More
TARGETMOL CHEMICALS INC SMI-16A 25MG
SDP
Catalog No. 502580029
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SMI-16a (PIM1/2 Kinase Inhibitor VI) a cell-permeable thiazolidinedione compound acts as an effective ATP-competitive inhibitor against Pim-1/2 kinases (IC50 150/20 nM) while exhibiting little or no activity against a panel of 57 other kinases (≤18% inhibition at 5 uM). purity: 99%
Catalog No. 50-258-0029 Supplier TARGETMOL CHEMICALS INC Supplier No. T398925MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SMI-16a (PIM1/2 Kinase Inhibitor VI) a cell-permeable thiazolidinedione compound acts as an effective ATP-competitive inhibitor against Pim-1/2 kinases (IC50 150/20 nM) while exhibiting little or no activity against a panel of 57 other kinases (≤18% inhibition at 5 uM). purity: 99%