Learn More
Medchemexpress LLC SMI-16a | 587852-28-6 | MFCD31382191 | 98.0% | 263.31 g·mol⁻¹ | C13H13NO3S | 1 ML
SDP
Catalog No. 5000249213
Encompass_Preferred
SMI-16a is a small-molecule research reagent that selectively inhibits Pim family kinases and is used to probe Pim-mediated signaling and cellular responses in vitro and in vivo. It shows potent biochemical activity against Pim1 and Pim2 and demonstrable cellular activity in prostate cancer cells.
Catalog No. 50-002-49213 Supplier Medchemexpress LLC Supplier No. HY10194710MM 1ML
SMI-16a is a small-molecule research reagent that selectively inhibits Pim family kinases and is used to probe Pim-mediated signaling and cellular responses in vitro and in vivo. It shows potent biochemical activity against Pim1 and Pim2 and demonstrable cellular activity in prostate cancer cells.