Learn More
Medchemexpress LLC HY-101947 5mg Medchemexpress, SMI-16a CAS:587852-28-6 Purity:>98%
SDP
Catalog No. 501967930
Encompass_Preferred
Medchemexpress, HY-101947 5mg SMI-16a CAS:587852-28-6 SMI-16a is a selective Pim kinase inhibitor with IC50 values of 0.15, 0.02 and 48 μM for Pim1, Pim2 and PC3 cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7930 Supplier Medchemexpress LLC Supplier No. HY1019475MG
Medchemexpress, HY-101947 5mg SMI-16a CAS:587852-28-6 SMI-16a is a selective Pim kinase inhibitor with IC50 values of 0.15, 0.02 and 48 μM for Pim1, Pim2 and PC3 cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.