Learn More
Selleck Chemical LLC SMI-4a
SDP
Supplier:  Selleck Chemical LLC S800510MG
Encompass_Preferred
SMI-4a (TCS PIM-1 4a) is a potent inhibitor of Pim1 with IC50 of 17 nM modestly potent to Pim-2 does not significantly inhibit any other serine/threonine- or tyrosine-kinases
Catalog No. 50-217-3335