Learn More
Apexbio Technology LLC SMI-4a 438190-29-5 50mg
SDP
Catalog No. 5000093622
Encompass_Preferred
SMI-4a (CAS 438190-29-5) is a selective inhibitor of the serine/threonine kinase Pim-1 exhibiting an IC50 of 17 nM Pim-1 is implicated in the regulation of cell cycle progression and apoptosis with its overexpression associated with enhanced cell survival in various malignancies SMI-4a demonstrates increased activity compared to related compounds and inhibits downstream mTORC1 signaling and activates AMPK in leukemia cell lines leading to cell cycle arrest and apoptosis induction In preclinical studies SMI-4a reduced tumor volume in xenograft mouse models highlighting its utility for investigating Pim kinase-driven pathways in cancer research
Catalog No. 50-000-93622 Supplier Apexbio Technology LLC Supplier No. A419350
SMI-4a (CAS 438190-29-5) is a selective inhibitor of the serine/threonine kinase Pim-1 exhibiting an IC50 of 17 nM Pim-1 is implicated in the regulation of cell cycle progression and apoptosis with its overexpression associated with enhanced cell survival in various malignancies SMI-4a demonstrates increased activity compared to related compounds and inhibits downstream mTORC1 signaling and activates AMPK in leukemia cell lines leading to cell cycle arrest and apoptosis induction In preclinical studies SMI-4a reduced tumor volume in xenograft mouse models highlighting its utility for investigating Pim kinase-driven pathways in cancer research