Learn More
Medchemexpress LLC SMI-4a | 438190-29-5 | 99.85% | 273.23 | 5 MG
SDP
Catalog No. 5000392164
Encompass_Preferred
SMI-4a (TCS-PIM-1-4a) is a potent, selective, cell-permeable, and ATP-competitive Pim-1 inhibitor with an IC50 of 24 μM and a Ki of 0.6 μM. It also inhibits Pim-2 (IC50 of 100 μM) but does not significantly inhibit other serine/threonine- or tyrosine-kinases. SMI-4a exhibits anticancer activity.
  • Potent, selective, cell-permeable, and ATP-competitive Pim-1 inhibitor.
  • Inhibits Pim-2.
  • Exhibits anticancer activity.
  • For research use only.
Catalog No. 50-003-92164 Supplier Medchemexpress LLC Supplier No. HY16576A5MG
Add to Cart
Add to Cart
SMI-4a (TCS-PIM-1-4a) is a potent, selective, cell-permeable, and ATP-competitive Pim-1 inhibitor with an IC50 of 24 μM and a Ki of 0.6 μM. It also inhibits Pim-2 (IC50 of 100 μM) but does not significantly inhibit other serine/threonine- or tyrosine-kinases. SMI-4a exhibits anticancer activity.
  • Potent, selective, cell-permeable, and ATP-competitive Pim-1 inhibitor.
  • Inhibits Pim-2.
  • Exhibits anticancer activity.
  • For research use only.