Learn More
Medchemexpress LLC SMI-4a | 438190-29-5 | 99.9% | 273.23 | 50 MG
SDP
Catalog No. 5000391714
Encompass_Preferred
SMI-4a (TCS-PIM-1-4a) is a potent, selective, cell-permeable, and ATP-competitive Pim-1 inhibitor. It inhibits Pim-1 with an IC50 of 24 μM and Pim-2 with an IC50 of 100 μM, and does not significantly inhibit other serine/threonine- or tyrosine-kinases. This compound exhibits anticancer activity and is supplied as a white to off-white solid.
  • Potent, selective, cell-permeable, and ATP-competitive Pim-1 inhibitor
  • Inhibits Pim-2
  • Exhibits anticancer activity
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO
Catalog No. 50-003-91714 Supplier Medchemexpress LLC Supplier No. HY16576A50MG
SMI-4a (TCS-PIM-1-4a) is a potent, selective, cell-permeable, and ATP-competitive Pim-1 inhibitor. It inhibits Pim-1 with an IC50 of 24 μM and Pim-2 with an IC50 of 100 μM, and does not significantly inhibit other serine/threonine- or tyrosine-kinases. This compound exhibits anticancer activity and is supplied as a white to off-white solid.
  • Potent, selective, cell-permeable, and ATP-competitive Pim-1 inhibitor
  • Inhibits Pim-2
  • Exhibits anticancer activity
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO