Learn More
AdipoGen SMI-4a (5 mg)
SDP
Supplier:  AdipoGen AGCR13503M005
Encompass_Preferred
Chemical. CAS: 327033-36-3. Formula: C11H6F3NO2S. MW: 273.2. Selective inhibitor of Pim-1 and Pim-2 protein kinases. Inducer of G1 phase cell cycle arrest. Inducer of p27Kip1. Inducer of apoptosis through the mitochondrial pathway. Inhibitor of the mammalian target of rapamycin C1 (mTORC1) pathway. Downregulates c-myc. Inhibitor of PRAS40 phosphorylation and mTOR activity. Potential anti-cancer compound. Blocks prostate cancer growth.
Catalog No. 50-205-3879