Learn More
Medchemexpress LLC Smifh2 | 340316-62-3 | 99.1% | 377.21 | 100 MG
SDP
Catalog No. 5000393843
Encompass_Preferred
SMIFH2 is a formin specific inhibitor that inhibits actin polymerization by Formins and affects the actin cytoskeleton. It induces dynamic cytoskeletal remodeling in U2OS cells and reduces p300, mDia2, and p53 levels through a proteasome-independent mechanism that influences remodeling of the cytoskeleton. This product is intended for research use only.
  • Inhibits actin polymerization
  • Affects the actin cytoskeleton
  • Induces dynamic cytoskeletal remodeling in U2OS cells
  • Reduces p300, mDia2, and p53 levels
  • Reduces p53 expression and activity through a post-transcriptional, proteasome-independent pathway
Catalog No. 50-003-93843 Supplier Medchemexpress LLC Supplier No. HY16931100MG
SMIFH2 is a formin specific inhibitor that inhibits actin polymerization by Formins and affects the actin cytoskeleton. It induces dynamic cytoskeletal remodeling in U2OS cells and reduces p300, mDia2, and p53 levels through a proteasome-independent mechanism that influences remodeling of the cytoskeleton. This product is intended for research use only.
  • Inhibits actin polymerization
  • Affects the actin cytoskeleton
  • Induces dynamic cytoskeletal remodeling in U2OS cells
  • Reduces p300, mDia2, and p53 levels
  • Reduces p53 expression and activity through a post-transcriptional, proteasome-independent pathway