Learn More
MSC Smith and Wesson Safety Glasses, Polycarbonate, Clear Mirror, 1 ea
Catalog No. 5000787236
Encompass
These safety glasses provide essential eye protection with a durable design. Featuring scratch-resistant polycarbonate lenses and a full-framed wrap-around style, they are suitable for various indoor and outdoor applications, meeting ANSI Z87.1+ safety standards.
  • Polycarbonate scratch-resistant clear mirror lenses
  • Full-framed wrap-around design
  • Black frame and temples
  • Non-adjustable temples
  • Provides UV protection
  • Meets ANSI Z87.1+ standards
  • Includes lens cleaning cloth and microfiber carry pouch
Catalog No. 50-007-87236 Supplier MSC Supplier No. 64304983
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These safety glasses provide essential eye protection with a durable design. Featuring scratch-resistant polycarbonate lenses and a full-framed wrap-around style, they are suitable for various indoor and outdoor applications, meeting ANSI Z87.1+ safety standards.
  • Polycarbonate scratch-resistant clear mirror lenses
  • Full-framed wrap-around design
  • Black frame and temples
  • Non-adjustable temples
  • Provides UV protection
  • Meets ANSI Z87.1+ standards
  • Includes lens cleaning cloth and microfiber carry pouch