Learn More
MSC Smith and Wesson Safety Glasses, Polycarbonate, Scratch-Resistant, 1 ea
Catalog No. 5000677557
Encompass
These safety glasses are designed with a full-framed, wrap-around style, featuring scratch-resistant, polycarbonate clear lenses for optimal eye protection. They meet ANSI Z87.1+ standards.
  • Scratch-resistant lens coating
  • Full-framed, wrap-around design
  • Clear polycarbonate lenses
  • Universal size for broad compatibility
  • Integrated nosepiece ridges to divert sweat
  • Provides UV protection
  • Meets ANSI Z87.1+ safety standards
Catalog No. 50-006-77557 Supplier MSC Supplier No. 00410449
These safety glasses are designed with a full-framed, wrap-around style, featuring scratch-resistant, polycarbonate clear lenses for optimal eye protection. They meet ANSI Z87.1+ standards.
  • Scratch-resistant lens coating
  • Full-framed, wrap-around design
  • Clear polycarbonate lenses
  • Universal size for broad compatibility
  • Integrated nosepiece ridges to divert sweat
  • Provides UV protection
  • Meets ANSI Z87.1+ safety standards