Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICU Medical Anaerobic Pulsator™ Plus Arterial Blood Sampling Kit

Catalog No. 22004195
Encompass
Change view
Click to view available options
Quantity:
100/Cs.
Needle Gauge:
22 G
Length (English) Needle:
1 in.
Product Type:
FP, NP devices; support kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Needle Gauge Length (English) Needle Product Type
22-004-195 100/Cs. 22 G 1 in. FP, NP devices; support kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-004-195 Supplier ICU Medical Supplier No. 4067NP1

missing translation for 'validMainframeMsg'

Pulsator™ Plus syringes, with a liquid sodium heparin concentration of 23.5 I.U. per mL, allowing blood sampling with minimal risk of sample dilution. The syringes use a specially formulated electrolyte-balanced liquid sodium heparin that offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels. The syringes are available with a broad choice of product features.

Anaerobic Pulsator Plus syringes, with a liquid sodium heparin concentration of 23.5 IU/mL, allow for clot-free blood sampling without risking sample dilution. Specially formulated electrolyte-balanced liquid sodium heparin offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels, providing consistent and accurate analysis for all parameters tested.

  • Consistent and accurate readings of pH, pOC2, pO2, Ca2+, Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, and lactateFill rapidly, to reduce fill time and minimize patient discomfort
  • Pulsating effect allows for precise positioning in arterial lumen
  • Syringe can be easily aspirated during a low blood pressure or code situation
  • Patented mini-volume insert needle and hub has a residual liquid heparin dead space of 0.03 mL; allows liquid heparin to be thoroughly expelled from syringe and needle, leaves behind an accurate trace coating of heparin
  • Needles available with different gauges, lengths and safety devices (Needle-Pro™, Point-Lok™ or Filter-Pro™)
  • With or without support kit (alcohol prep pad, iodophor prep pad, two gauze pads, patient label, biohazard labeled ice bag, adhesive bandage)

Order Info

  • Needles available with different gauges, lengths and safety devices (Needle-Pro™, Point-Lok™ or Filter-Pro™)
  • With or without support kit (alcohol prep pad, iodophor prep pad, two gauze pads, patient label, biohazard labeled ice bag, adhesive bandage)

Specifications

Needle Gauge 22 G
For Use With (Application) Allow for clot-free blood sampling without risking sample dilution, offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels, providing consistent and accurate analysis for all parameters tested
Includes 3mL luer-lock syringe with Filter-Pro™ device, 22G x 1 in. needle with separate Needle-Pro™ needle safety device, mini-volume insert needle, accessory kit
Length (English) Needle 1 in.
Product Type FP, NP devices; support kit
Quantity 100/Cs.
Packaging Polybag
Volume (Metric) 3 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.