Learn More
Smiths Medical Hypodermic Needle-Pro™ Device and Syringe
Catalog No. 22004261
Change view
Click to view available options
Syringe inc.; Needle hub: Orange; 25G x 5/8 in.
Catalog No.
22-004-261 Syringe inc.; Needle hub: Orange; 25G x 5/8 in.
1 options
This item is not returnable. View return policy
Catalog No. 22-004-261 Supplier Smiths Medical Supplier No. ACC000038
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rotating safety device facilitates intradermal injections and does not contribute to additional dead space to standard needle/syringe injections
  • Meets NIOSH/CDC recommendations for safety devices
  • Safety feature engages with a single-handed technique
  • Facilitates two-needle injection techniques