Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICU Medical Portex™ Line Draw Plus Arterial Blood Sampling Kits

Catalog No. 22004196
Encompass
Change view
Click to view available options
Quantity:
200/Cs.
Volume (Metric):
1 mL
3 mL
Packaging:
Polybag
Polybag and Biohaz Bag
Product Type:
Anaerobic Na Heparin LD Syr
Li Heparin Syringe, Waste Syr
Lithium Heparin LL Syringe
Lithium Heparin LS Syringe
Low Lithium Heparin LL Syringe
Low Lithium Heparin LS Syringe
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Volume (Metric) Packaging Product Type
22-004-196 200/Cs. 3 mL Polybag Lithium Heparin LS Syringe
22-004-197 200/Cs. 1 mL Polybag Lithium Heparin LS Syringe
22-024-978 200/Cs. 1 mL Polybag Low Lithium Heparin LS Syringe
22-024-957 200/Cs. 1 mL Polybag Li Heparin Syringe, Waste Syr
22-024-987 200/Cs. 3 mL Polybag Lithium Heparin LL Syringe
22-024-958 200/Cs. 3 mL Polybag Low Lithium Heparin LL Syringe
22-024-960 200/Cs. 3 mL Polybag and Biohaz Bag Lithium Heparin LS Syringe
22-024-966 200/Cs. 3 mL Polybag Anaerobic Na Heparin LD Syr
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 22-004-196 Supplier ICU Medical Supplier No. 40432
Add to Cart
Add to Cart

Line Draw Plus aspirating syringes allow for blood sampling directly from an arterial line. The syringes use an advanced formula of dry lithium heparin for consistent and accurate readings. They are available with the following configurations: 1 mL Luer Slip Syringe, 3 mL Luer Lock Syringe, or 3 mL Luer Slip Slip Syringe.

Advanced formula offsets both binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level. Provides consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, and lactate.

  • Umbilical and arterial syringes contain a 23.5 IU/mL concentration of dry lithium heparin
  • Low-heparin arterial syringes have a 7 IU/mL concentration of dry lithium heparin
  • Anaerobic syringes contain 23.5 IU/mL of electrolyte-balanced liquid sodium heparin

†With support kit (alcohol prep pad, gauze pad, biohazard-labeled ice bag).

Specifications

For Use With (Application) Allow for blood sampling directly from an arterial line, provides consistent and accurate analysis for all parameters tested
Includes 3mL line draw luer-slip syringe with Filter-Pro™ device, 23.5 IU/mL concentration of dry lithium heparin as anticoagulant
Product Type Lithium Heparin LS Syringe
Quantity 200/Cs.
Packaging Polybag
Volume (Metric) 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.