Learn More
ICU Medical Portex™ Line Draw Plus Arterial Blood Sampling Kits
Description
Advanced formula offsets both binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level. Provides consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, and lactate.
- Umbilical and arterial syringes contain a 23.5 IU/mL concentration of dry lithium heparin
- Low-heparin arterial syringes have a 7 IU/mL concentration of dry lithium heparin
- Anaerobic syringes contain 23.5 IU/mL of electrolyte-balanced liquid sodium heparin
†With support kit (alcohol prep pad, gauze pad, biohazard-labeled ice bag).
Specifications
Specifications
| Includes | Anaerobic Line Draw-Liquid Sodium Heparin; 3mL Luer slip; Filter-Pro; Polybag packaging |
| Product Type | Anaerobic Na Heparin LD Syr |
| Quantity | 200/Cs. |
| Packaging | Polybag |
| Volume (Metric) | 3 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.