Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICU Medical Portex™ Line Draw Plus Arterial Blood Sampling Kits
Click to view available options
Quantity:
200/Cs.
Volume (Metric):
1 mL
3 mL
Packaging:
Polybag
Polybag and Biohaz Bag
Product Type:
Anaerobic Na Heparin LD Syr
Li Heparin Syringe, Waste Syr
Lithium Heparin LL Syringe
Lithium Heparin LS Syringe
Low Lithium Heparin LL Syringe
Low Lithium Heparin LS Syringe
Description
Advanced formula offsets both binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level. Provides consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, and lactate.
- Umbilical and arterial syringes contain a 23.5 IU/mL concentration of dry lithium heparin
- Low-heparin arterial syringes have a 7 IU/mL concentration of dry lithium heparin
- Anaerobic syringes contain 23.5 IU/mL of electrolyte-balanced liquid sodium heparin
†With support kit (alcohol prep pad, gauze pad, biohazard-labeled ice bag).
Specifications
Specifications
| For Use With (Application) | Allow for blood sampling directly from an arterial line, provides consistent and accurate analysis for all parameters tested |
| Includes | 3mL line draw luer-lock syringe with Filter-Pro™ device, 23.5 IU/mL concentration of dry lithium heparin as anticoagulant |
| Product Type | Lithium Heparin LL Syringe |
| Quantity | 200/Cs. |
| Packaging | Polybag |
| Volume (Metric) | 3 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction