Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICU Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 3cc/70.5IU Kits

Catalog No. 22004190
Encompass
Change view
Click to view available options
Quantity:
100/Cs.
200/Cs.
Product Type:
Arterial Blood Sampling Kit
FP, NP LL, 2nd needle, kit, pp
FP, NP LS device, kit, pp
FP, NP LS device, support kit
FP, NP LS, pad, label, ice bag
FP, PL LL device, kit, pp
FP, PL LL, 2nd needle, kit, pp
FP,NP LS, 2nd needle, kit, pp
Filter-Pro, Needle-Pro LL dev
Filter-Pro, Needle-Pro LS dev
Filter-Pro, Point-Lok LL dev
Needle Gauge:
22 G
22 to 23 G
23 G
Packaging:
Sterile Tyvek
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Needle Gauge Packaging
22-004-190 100/Cs. FP, PL LL, 2nd needle, kit, pp 22 to 23 G Sterile Tyvek
22-004-189 100/Cs. FP, PL LL device, kit, pp 22 G Sterile Tyvek
22-024-992 200/Cs. Filter-Pro, Needle-Pro LL dev 23 G Sterile Tyvek
22-024-990 200/Cs. Filter-Pro, Point-Lok LL dev 23 G Sterile Tyvek
22-024-991 200/Cs. Filter-Pro, Point-Lok LL dev 22 G Sterile Tyvek
22-004-188 100/Cs. FP, NP LL, 2nd needle, kit, pp 22 to 23 G Sterile Tyvek
22-004-183 200/Cs. Filter-Pro, Needle-Pro LS dev 23 G Sterile Tyvek
22-024-980 100/Cs. FP, NP LS device, kit, pp 22 G Sterile Tyvek
22-004-181 100/Cs. FP, NP LS, pad, label, ice bag 22 G Sterile Tyvek
22-004-180 100/Cs. FP, NP LS device, kit, pp 23 G Sterile Tyvek
22-024-952 100/Cs. FP, NP LS device, support kit 23 G Sterile Tyvek
22-004-179 200/Cs. FP,NP LS, 2nd needle, kit, pp 23 G Sterile Tyvek
22-004-178 100/Cs. Arterial Blood Sampling Kit 22 to 23 G Sterile Tyvek
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 22-004-190 Supplier ICU Medical Supplier No. 4589P1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Only available in the USA and selectively in Canada. Pro-Vent™ Plus 3 mL Arterial Blood Sampling syringes utilize lithium heparin with a concentration of 23.5 I.U. per mL. The specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference for more accurate and consistent results. The syringes are available with a broad choice of product features.

  • Advanced-formula dry lithium heparin
  • Offsets binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level
  • Consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, lactate
  • Large venting filter media facilitates quick filling and the siliconized barrel allows easy aspiration, even in code situations
  • Filter-Pro™ device permits ease of removing air bubbles without accidentally expelling blood or aerosolizing the sample
  • Needle-Pro™ needle protection device (sturdy, puncture-resistant material) lock in place, completely covering used needle
  • Support kits contain alcohol prep pad, lodophor prep pad, gauze pad, patient label, biohazard-labeled ice bag, adhesive bandage

Order Info

Sterile Tyvek packaging except as noted.

†Polybagged.

Specifications

Needle Gauge 22 to 23 G
For Use With (Application) Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results
Includes 3mL luer-lock syringe with Filter-Pro™ device, 23G x 1 in. preattached needle, 22G x 1.5 in. secondary needle, Point-Lok™ device, support kit
Product Type FP, PL LL, 2nd needle, kit, pp
Quantity 100/Cs.
Packaging Sterile Tyvek
Volume (Metric) 3 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.