Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICU Medical Pro-Vent™ Plus Syringes with Dry Lithium Heparin

Catalog No. 22024989
Encompass
Change view
Click to view available options
Needle Gauge:
23 G
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
22-024-989 23 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-024-989 Supplier ICU Medical Supplier No. 4629P2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pro-Vent™ Plus 1 mL Arterial Blood Sampling syringes utilize lithium heparin with a concentration of 23.5 I.U. per mL. The specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference for more accurate and consistent results.The syringes are available with a broad choice of product features.

  • Needles with different gauges and lengths
  • Needle safety devices (Needle-Pro™ and Point-Lok™)
  • Support kit (alcohol prep pad, gauze pad, patient label, biohazard labeled ice bag, adhesive bandage)
  • Iodophor prep pad

Specifications

Needle Gauge 23 G
Length (English) Needle 1 in.
Product Type Syringe
Volume (Metric) 1 mL
Connection Type Leur Slip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.