Learn More
ICU Medical Pulsator™ Plus Dry Heparin Arterial Blood Sampling Kits
Catalog No. 22024976 Shop All Smiths Medical PM Inc. Products
Change view
Click to view available options
Quantity:
100/Cs.
200/Cs.
Product Type:
With 2nd Needle, Accessory Kit
With Accessory Kit
With label, adhesive bandage
Without Needle
Packaging:
Polybag
Sterile Tyvek™
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Packaging
22-024-976 200/Cs. Without Needle Polybag
22-024-988 200/Cs. With label, adhesive bandage Sterile Tyvek™
22-024-949 100/Cs. With 2nd Needle, Accessory Kit Sterile Tyvek™
22-024-975 100/Cs. With Accessory Kit Sterile Tyvek™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-024-976 Supplier ICU Medical Supplier No. 40462
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
3-cc or 5-cc Luer lock syringe and Filter-Pro™ device in polybag or Tyvek packaging.
Pulsator™ Plus 3 mL arterial blood sampling syringes utilize lithium heparin with a concentration of 23.5 I.U. per mL. The specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference with electrolyte values, for more accurate and consistent results. The syringes are available with a choice of product features.
  • Pulsating effect allows for precise positioning in arterial lumen
  • Syringe response to arterial pressure helps ensure arterial sample
  • Rapid fill minimizes patient discomfort
  • 3mL arterial blood-sampling syringes are easily aspirated during low blood pressure or code situation
  • Needles with different gauges and lengths
  • Utilize lithium heparin with a concentration of 23.5 IU/mL
  • Specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference for more accurate and consistent results
Order Info
  • With or without support kit (alcohol prep pad, iodophor prep pad, gauze pad, patient label, biohazard labeled ice bag, adhesive bandage)
  • Sterile Tyvek™ packaging
  • Come with Filter-Pro™ air bubble removal device
†With support kit.
Specifications
For Use With (Application) Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results, rapid fill minimizes patient discomfort
Includes 3mL luer-lock syringe with Filter-Pro™ air bubble removal device, 23.5 IU/mL concentration of dry lithium heparin as anticoagulant
Product Type Without Needle
Quantity 200/Cs.
Packaging Polybag
Volume (Metric) 3 mL