Learn More
ICU Medical Jelco™ Saf-T Holder™ Blood Tube Holders
Catalog No. 22024845
Encompass
Change view
Click to view available options
Connectors:
Female Luer
Male Luer
Product Type:
Blood Collection Tube Holder
Sterility:
Sterile
For Use With (Equipment):
Saf-T Wing, Saf-T Close
Trek Diagnostic Systems
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connectors Product Type Sterility For Use With (Equipment)
22-024-845 Female Luer Blood Collection Tube Holder Sterile Saf-T Wing, Saf-T Close
22-024-840 Male Luer Blood Collection Tube Holder Sterile Saf-T Wing, Saf-T Close
22-024-826 Male Luer Blood Collection Tube Holder Sterile Trek Diagnostic Systems
22-024-827 Female Luer Blood Collection Tube Holder Sterile Trek Diagnostic Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-024-845 Supplier ICU Medical Supplier No. 96002
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Saf-T Holder™ devices come with the back-end needle and multi-sample Luer permanently attached to the holder to minimize potential exposures and holder re-use. Available separately or with Saf-T Wing™ Blood Collection & Infusion Sets, Saf-T Holder™ devices also provide easy needless transfer after a syringe draw.
For use with Saf-T Wing™ winged infusion sets and Saf-T Closed Blood Collection System™ sets.
  • Back-end needle and multi-sample Luer permanently attached to the holder to eliminate potential exposures and holder re-use
  • Available with male or female Luer adapter
  • Sterilized with ethylene oxide
  • Latex-free
Order Info
Standard length holder fits most blood collection systems. Shorter length designed for use exclusively with Trek Diagnostic systems.
Certifications
CE marked
Specifications
For Use With (Equipment) Saf-T Wing, Saf-T Close
Sterility Sterile
Connectors Female Luer
Product Type Blood Collection Tube Holder
Certifications/Compliance CE, OSHA
Latex Free Yes
For Use With (Application) Provide safe needleless transfer after a syringe draw