Learn More
ICU Medical Venipuncture Needle-Pro™ Device
Catalog No. 22004141
Encompass
Change view
Click to view available options
Includes:
1000 Devices, Bulk Pack
1000 Devices, Individual Bags
500 Devices, Dispenser Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-004-141 1000 Devices, Bulk Pack
22-004-140 1000 Devices, Individual Bags
22-004-142 500 Devices, Dispenser Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-004-141 Supplier ICU Medical Supplier No. 4141
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
The Venipuncture Needle-Pro™ device is designed to protect clinicians from exposure to contaminated blood collection needles. The holder-based safety device provides protection on both ends and prevents holder re-use, assuring compliance with OSHA directives.
  • Disposable holder to eliminate cross-contamination
  • One-piece holder
  • Single-handed engagement
  • Integral needle protection device that can be used with any needle and vacuum tube system
  • Swiveling needle sheath allows clinician to locate the needle bevel in the desired position throughout the procedure
  • Disposable holder reduces the risk of reusing contaminated needle holders and provides end-to-end protection
Specifications
Includes 1000 Devices, Bulk Pack
Certifications/Compliance NIOSH CDC, OSHA
Product Type Needle Protection Device
For Use With (Equipment) All needle and vacuum tube system
For Use With (Application) Quick and effective means of needlestick protection for phlebotomists, nurses and other clinicians