Learn More
ICU Medical Venipuncture Needle-Pro™ Device
Catalog No. p-177785
Encompass
Change view
Click to view available options
Includes:
1000 Devices, Bulk Pack
1000 Devices, Individual Bags
500 Devices, Dispenser Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-004-140 1000 Devices, Individual Bags
22-004-141 1000 Devices, Bulk Pack
22-004-142 500 Devices, Dispenser Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-004-140 Supplier ICU Medical Supplier No. 4140
Only null left
Add to Cart
Edge
Add to Cart
The Venipuncture Needle-Pro™ device is designed to protect clinicians from exposure to contaminated blood collection needles. The holder-based safety device provides protection on both ends and prevents holder re-use, assuring compliance with OSHA directives.
  • Disposable holder to eliminate cross-contamination
  • One-piece holder
  • Single-handed engagement
  • Integral needle protection device that can be used with any needle and vacuum tube system
  • Swiveling needle sheath allows clinician to locate the needle bevel in the desired position throughout the procedure
  • Disposable holder reduces the risk of reusing contaminated needle holders and provides end-to-end protection
Specifications
Includes 1000 Devices, Individual Bags
For Use With (Equipment) All needle and vacuum tube system
Certifications/Compliance NIOSH CDC, OSHA
Product Type Needle Protection Device
For Use With (Application) Quick and effective means of needlestick protection for phlebotomists, nurses and other clinicians