Learn More
Electron Microscopy Sciences Small SEM Clamp, 15 x 10 mm, 3.2 mm
SDP
Catalog No. 501927143
Encompass_Preferred
15 mm (0.6") long with 10 mm (0.4") wide clamping area and standard pin stub 3.2 mm ( 1/8"). Suitable for holding samples up to 7 mm overall thickness. Perfect for small tubes, strips, wires, and smaller flat samples. Overall Dimensions 23 x 20 x 9 mm (0.9" x 0.78" x 0.35"). Made from machined aluminum with brass screws.
Size: 15 x 10 mm
Pin diameter: 3.2 mm
Catalog No. 50-192-7143 Supplier Electron Microscopy Sciences Supplier No. 7595402
15 mm (0.6") long with 10 mm (0.4") wide clamping area and standard pin stub 3.2 mm ( 1/8"). Suitable for holding samples up to 7 mm overall thickness. Perfect for small tubes, strips, wires, and smaller flat samples. Overall Dimensions 23 x 20 x 9 mm (0.9" x 0.78" x 0.35"). Made from machined aluminum with brass screws.
Size: 15 x 10 mm
Pin diameter: 3.2 mm