Learn More
New England Biolabs, Inc. SmlI – 500 units
SDP
Catalog No. 50812316
Encompass
A restriction endonuclease that recognizes the sequence C^TYRA_G.
Catalog No. 50-812-316 Supplier New England Biolabs, Inc. Supplier No. R0597S
A restriction endonuclease that recognizes the sequence C^TYRA_G.