Learn More
Sino Biological SMM 293-TII Expression Medium (Serum free, complete medium)
Catalog No. NC2750985
Encompass_Preferred
SMM 293-TII Expression Medium is an optimized, animal origin components-free medium used to support the growth and transfection of HEK293 cells in suspension conditions. This medium is complete and ready-to-use, which does not require serum and glutamine as supplementations. SMM 293-TII Expression Medium performs excellent in the high cell density culture and effective expression of target molecules. SMS 293-SUPISupplementations (Catalog No. M293-SUPI) are recommended in expression phase for long-term culture and higher yield.
Catalog No. NC2750985 Supplier Sino Biological Supplier No. M293TII
SMM 293-TII Expression Medium is an optimized, animal origin components-free medium used to support the growth and transfection of HEK293 cells in suspension conditions. This medium is complete and ready-to-use, which does not require serum and glutamine as supplementations. SMM 293-TII Expression Medium performs excellent in the high cell density culture and effective expression of target molecules. SMS 293-SUPISupplementations (Catalog No. M293-SUPI) are recommended in expression phase for long-term culture and higher yield.