Learn More
Carnabio Usa Inc SMMLCK(MYLK)/10UG/CATALYTIC
Supplier:  Carnabio Usa Inc 0215110
Encompass
smMLCK(MYLK), Catalytic domain, Wild type, Amino Acid 1428-1771, NP_444253.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0988