Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Smn-c2 | 1446311-56-3 | 98.9% | 417.50 | C24H27N5O2 | 10MM 1ML
SDP

Catalog No. 5000237149
Encompass_Preferred

SMN-C2 is a small-molecule SMN2 splicing regulator (an analog of RG-7916) used in spinal muscular atrophy research. It modulates SMN2 pre-mRNA splicing by increasing the affinity of RNA-binding proteins FUBP1 and KHSRP for the SMN2 pre-mRNA complex.

  • Small-molecule SMN2 splicing regulator for research use only.
  • Modulates SMN2 pre-mRNA splicing by increasing FUBP1 and KHSRP binding.
  • Available as 10 mM solution in DMSO (1 mL) and as solid in multiple mg sizes.
  • Purity 98.9%.
  • Recommended storage: powder -20°C (up to 3 years); in solvent -80°C (up to 6 months).

Catalog No. 50-002-37149 Supplier Medchemexpress LLC Supplier No. HY12464810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SMN-C2 is a small-molecule SMN2 splicing regulator (an analog of RG-7916) used in spinal muscular atrophy research. It modulates SMN2 pre-mRNA splicing by increasing the affinity of RNA-binding proteins FUBP1 and KHSRP for the SMN2 pre-mRNA complex.

  • Small-molecule SMN2 splicing regulator for research use only.
  • Modulates SMN2 pre-mRNA splicing by increasing FUBP1 and KHSRP binding.
  • Available as 10 mM solution in DMSO (1 mL) and as solid in multiple mg sizes.
  • Purity 98.9%.
  • Recommended storage: powder -20°C (up to 3 years); in solvent -80°C (up to 6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.