Learn More
Medchemexpress LLC SMN-C2 | 1446311-56-3 | 98.9% | 417.50 g/mol | C24H27N5O2 | 100MG
SDP
Catalog No. 5000237924
Encompass_Preferred
SMN-C2 is a small-molecule modulator of SMN2 pre-mRNA splicing used in research to promote SMN2 exon inclusion. It is an analog of RG-7916 and is characterized for use in cellular and biochemical assays investigating splicing modulation and spinal muscular atrophy pathways.
  • High purity suitable for research use (98.9%).
  • Modulates SMN2 splicing to increase exon inclusion.
  • Chemical formula C24H27N5O2; molecular weight 417.50 g/mol.
  • Available in multiple milligram quantities, including 100 mg.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months).
Catalog No. 50-002-37924 Supplier Medchemexpress LLC Supplier No. HY124648100MG
SMN-C2 is a small-molecule modulator of SMN2 pre-mRNA splicing used in research to promote SMN2 exon inclusion. It is an analog of RG-7916 and is characterized for use in cellular and biochemical assays investigating splicing modulation and spinal muscular atrophy pathways.
  • High purity suitable for research use (98.9%).
  • Modulates SMN2 splicing to increase exon inclusion.
  • Chemical formula C24H27N5O2; molecular weight 417.50 g/mol.
  • Available in multiple milligram quantities, including 100 mg.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months).