Learn More
Medchemexpress LLC HY-112633 10mg Medchemexpress, SMN-C3 CAS:1449597-34-5 Purity:>98%
SDP
Catalog No. 501969124
Encompass_Preferred
Medchemexpress, HY-112633 10mg SMN-C3 CAS:1449597-34-5 SMN-C3 is an orally active SMN2 splicing modulator and has the potential to treat spinal muscular atrophy (SMA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9124 Supplier Medchemexpress LLC Supplier No. HY11263310MG
Medchemexpress, HY-112633 10mg SMN-C3 CAS:1449597-34-5 SMN-C3 is an orally active SMN2 splicing modulator and has the potential to treat spinal muscular atrophy (SMA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.