Learn More
TARGETMOL CHEMICALS INC SMN-C3 5MG
SDP
Catalog No. 502581393
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. SMN-C3 (MV8T2MCK57) is an orally active modulator of SMN2 splicing and has the potential to treat spinal muscular atrophy (SMA). purity: 98%
Catalog No. 50-258-1393 Supplier TARGETMOL CHEMICALS INC Supplier No. T129355MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. SMN-C3 (MV8T2MCK57) is an orally active modulator of SMN2 splicing and has the potential to treat spinal muscular atrophy (SMA). purity: 98%