Learn More
Enzo Life Sciences SMN1 monoclonal antibody (2B1) (200µg)
SDP
Supplier:  Enzo Life Sciences ADINBA202200
Encompass_Preferred
Clone: 2B1. Host: Mouse. Isotype: IgG. Immunogen: Recombinant fragment corresponding to the carboxy-terminus of human SMN1. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Applications: WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7358