Learn More
Aviva Systems Biology Corp SMNDC1 Antibody - middle region (ARP40612_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40612_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMNDC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SMNDC1. Alternate names: SMNR, SPF30ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57546