Learn More
AdipoGen SMNP mAb 7B10
SDP
Supplier:  AdipoGen AG20T0003C200
Encompass_Preferred
Monoclonal Antibody. Recognizes a sequence of up to 9 amino acids in the N-terminal domain of human SMN. Isotype Mouse IgG1. Clone 7B10. Applications IP, WB. Liquid. In PBS. The SMN complex plays an essential role in spliceosomal snRNP assembly in the cytoplasm and is required for pre-mRNA splicing in the nucleus. It may also play a role in the metabolism of snoRNPs. It is expressed in a wide variety of tissues, including high levels in brain, kidney and liver, moderate levels in skeletal and cardiac muscle and low levels in fibroblasts and lymphocytes. Mutations in SMN cause spinal muscular atrophy 1 SMA1. Spinal muscular atrophy is a group of neuromuscular disorders characterized by degeneration of the anterior horn cells of the spinal cord, leading to symmetrical muscle weakness and atrophy. The antibody was used extensively to purify and characterize the macromolecular SMN complex from cultured human cells.
Catalog No. 50-256-3361