Learn More
Sheldon Manufacturing Inc Oven, Forced Air, 1.5 cuft, 115v
SDP
Catalog No. NC1655806
Encompass_Preferred
The SMO1 has a quick ramp-up time to 306°C for faster cycle times. This unit includes an advanced PID temperature control system for sensitive response. These ovens are recommended for critical and analytical use in laboratory and pilot plant product research. Typical applications for the oven include aging, asphalt testing, curing, drying, pre-heating, conditioning, and life cycle testing. This unit is equipped with a stainless-steel interior for long-life operation and easy cleaning. Its triple wall construction minimizes external wall temperature which meets CE requirements to keep workspaces cool. The SMO1's forced air ovens deliver precise uniformity, air distribution and the peace of mind expected of the SHEL LAB brand.
Catalog No. NC1655806 Supplier Sheldon Manufacturing Inc Supplier No. SMO1
The SMO1 has a quick ramp-up time to 306°C for faster cycle times. This unit includes an advanced PID temperature control system for sensitive response. These ovens are recommended for critical and analytical use in laboratory and pilot plant product research. Typical applications for the oven include aging, asphalt testing, curing, drying, pre-heating, conditioning, and life cycle testing. This unit is equipped with a stainless-steel interior for long-life operation and easy cleaning. Its triple wall construction minimizes external wall temperature which meets CE requirements to keep workspaces cool. The SMO1's forced air ovens deliver precise uniformity, air distribution and the peace of mind expected of the SHEL LAB brand.