Learn More
MSC SDI Smoke Detector Tester, Smoke Centurion, Universal, 2.6 oz, 12/Case
Catalog No. NC1472809
Encompass
The Smoke Centurion Smoke Detector Tester is an aerosol tester designed to check the functionality of smoke detectors. It offers a fast and easy-to-use method for testing, suitable for a wide range of detectors, and can be used either handheld or with a dispenser. Its silicone-free formula helps reduce the risk of detector contamination.
  • More tests per can
  • Fast detector activation
  • Lowest cost per test
  • Flexible usage (handheld or with a dispenser)
  • Silicone-free testing reduces risk of detector contamination
  • Formula approved by leading detector manufacturers
  • Ozone friendly and low GWP propellant
Catalog No. NC1472809 Supplier MSC Supplier No. 57466047
The Smoke Centurion Smoke Detector Tester is an aerosol tester designed to check the functionality of smoke detectors. It offers a fast and easy-to-use method for testing, suitable for a wide range of detectors, and can be used either handheld or with a dispenser. Its silicone-free formula helps reduce the risk of detector contamination.
  • More tests per can
  • Fast detector activation
  • Lowest cost per test
  • Flexible usage (handheld or with a dispenser)
  • Silicone-free testing reduces risk of detector contamination
  • Formula approved by leading detector manufacturers
  • Ozone friendly and low GWP propellant