Learn More
Neuromics, Inc SMOOTH COAT SOLUTION,20ML
Catalog No. NC2298940
Encompass
CATALOG NUMBER: SC300APPLICATIONS: Cell AssaysTYPE: Cell Growth MediaSTORAGE: Store at 4°C short term. Store at -20°C long term.SHIPPING: Frozen (Polar Packs)FORMAT A: liquidSPECIES REACTIVITY: Human
Catalog No. NC2298940 Supplier Neuromics, Inc Supplier No. SC300
CATALOG NUMBER: SC300APPLICATIONS: Cell AssaysTYPE: Cell Growth MediaSTORAGE: Store at 4°C short term. Store at -20°C long term.SHIPPING: Frozen (Polar Packs)FORMAT A: liquidSPECIES REACTIVITY: Human