Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Smooth Disposable CPE Gown
GREENER_CHOICE

Catalog No. 12893010 Shop All Fisherbrand Products
Change view
Click to view available options
Autoclavable:
Not Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
12-893-010 Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-893-010
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides light-weight, impervious protection against bio-hazards, with non-linting construction for greater comfort.

  • In compliance with FDA requirements and tested in accordance with ASTM standards, including breaking load, tensile strength, and elongation
  • Successfully tested for liquid and aerosol blood resistance under ASTM F 1670, synthetic blood penetration and resistance, and ASTM F 1671 bacteriophage penetration resistance
  • Superior physical properties over imports
  • XL size with easy, tear-away feature for quick gown removal and safe disposal
  • Contoured thumb loops secure gown sleeve for added protection
  • Redirects stress to the film material not the thermal seals to minimize breakage potential
  • Full-length open back affords better comfort and shoulder coverage
  • Provides liquid and aerosol blood resistance
  • Lightweight, non-linting construction

Specifications

Size X-Large
Width (Metric) 200 cm
For Use With (Application) Industrial, Food Processing, Critical Environments, Healthcare
Gender Unisex
Length (Metric) 122 cm
Product Type Isolation Gown
No. of Pockets None
Material Cellulose, Polyester
Color Blue
Closure Type Waist Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Finish Smooth Matte
Sleeve Style Full Sleeve with Thumb Loop
Certifications/Compliance ASTM F 1670, ASTM 1671
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.