Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Smooth Muscle Growth Supplement (SMGS)

Catalog No. S00725
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00725 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00725 Supplier Gibco™ Supplier No. S00725
  • $161.00
    $152.65 / Each of 1

    Save $8.35

    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Smooth Muscle Growth Supplement (SMGS)

A sterile, concentrated (20X) solution intended for use with Medium 231 to culture human smooth muscle cells. Each 25 ml bottle of SMGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of human smooth muscle cells, and is the correct amount of supplement for a 500 ml bottle of basal Medium 231.

When a 500 ml bottle of medium is supplemented with SMGS, the final concentrations of the components in the supplemented medium are:

  • Fetal bovine serum (4.9% v/v)
  • Human basic fibroblast growth factor (2 ng/ml)
  • Human epidermal growth factor (0.5 ng/ml)
  • Heparin (5 ng/ml)
  • Recombinant human insulin-like growth factor-I (2μg/ml)
  • BSA (0.2μg/ml)
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store in freezer (-5 to -20°C).
    Cell Type Smooth Muscle Cells
    Concentration 20 X
    Product Type Growth Supplement
    Sterility Sterile
    Product Line Cascade Biologics
    Quantity 25 mL
    Shipping Condition Dry Ice
    Species Human
    WARNING: Cancer - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.