Learn More
Biocare Medical, LLC SMOOTH MUSCLE MYOSIN HC 0.5ML
SDP
Supplier:  Biocare Medical, LLC CM420B
Encompass_Preferred
Smooth Muscle Myosin Heavy Chain stains the intact myoepithelial cells present in breast lesions and bronchioloalveolar tissues. It can help distinguish benign from malignant and also reacts with visceral and vascular smooth muscle cells.
Catalog No. 50-827-12