Learn More
Research Products International Corp Smooth Rack, Poly, Blue, 72 Tubes
SDP
Catalog No. 501978551
Encompass
Innovative rack features uniquely designed supports that prevent tubes from slipping out of position. Holds a variety of tube sizes from 10 - 17 mm diameter with a total capacity of 72 tubes. Manufactured from heavy duty polypropylene for durability and excellent chemical resistance. Rack can be autoclaved.
Catalog No. 50-197-8551 Supplier Research Products International Corp Supplier No. 182021
Innovative rack features uniquely designed supports that prevent tubes from slipping out of position. Holds a variety of tube sizes from 10 - 17 mm diameter with a total capacity of 72 tubes. Manufactured from heavy duty polypropylene for durability and excellent chemical resistance. Rack can be autoclaved.