Learn More
Smooth On Inc SMOOTH-SIL 945 2LBS
SDP
Catalog No. NC2008255
Encompass
Platinum Silicones cure at room temperature with negligible shrinkage. These silicones exhibit good chemical, abrasion and heat resistance. Materials such as plasters, concrete, wax, low-melt metal alloys or resins (urethane, epoxy or polyester) can then be cast into these silicone rubbers without a release agent. Smooth-Sil 945 offers the convenience of a 1A:1B by volume mix ratio and a fast 6 hour cure time. Platinum Silicones are used for rapid prototyping, wax casting (foundries and candle makers), architectural restoration and for casting concrete.
Catalog No. NC2008255 Supplier Smooth On Inc Supplier No. 37041
Platinum Silicones cure at room temperature with negligible shrinkage. These silicones exhibit good chemical, abrasion and heat resistance. Materials such as plasters, concrete, wax, low-melt metal alloys or resins (urethane, epoxy or polyester) can then be cast into these silicone rubbers without a release agent. Smooth-Sil 945 offers the convenience of a 1A:1B by volume mix ratio and a fast 6 hour cure time. Platinum Silicones are used for rapid prototyping, wax casting (foundries and candle makers), architectural restoration and for casting concrete.