Learn More
Smooth On Inc SMOOTH-SIL 950 2.2LBS
SDP
Catalog No. NC2008248
Encompass
Platinum Silicones cure at room temperature with negligible shrinkage. These silicones exhibit good chemical, abrasion and heat resistance. Materials such as plasters, concrete, wax, low-melt metal alloys or resins (urethane, epoxy or polyester) can then be cast into these silicone rubbers without a release agent. Smooth-Sil Platinum Silicones are used for rapid prototyping, wax casting (foundries and candle makers), architectural restoration and for casting concrete. Smooth-Sil 950 is suitable for food related applications.
Catalog No. NC2008248 Supplier Smooth On Inc Supplier No. 83541
Platinum Silicones cure at room temperature with negligible shrinkage. These silicones exhibit good chemical, abrasion and heat resistance. Materials such as plasters, concrete, wax, low-melt metal alloys or resins (urethane, epoxy or polyester) can then be cast into these silicone rubbers without a release agent. Smooth-Sil Platinum Silicones are used for rapid prototyping, wax casting (foundries and candle makers), architectural restoration and for casting concrete. Smooth-Sil 950 is suitable for food related applications.