Learn More
Selleck Chemical LLC Smoothened Agonist SAG-5mg
SDP
Supplier:  Selleck Chemical LLC S63845MG
Encompass_Preferred
Smoothened Agonist (SAG) is a potent agonist of Smoothened (Smo). Smoothened Agonist induces firefly luciferase expression in Shh-LIGHT2 cells with an EC50 of 3 nM.
Catalog No. 50-238-9982