Learn More
Alzet iPRECIO Micro Infusion Pumps SMP-310R: Implantable, Programmable, Refillable Infusion Device for Mice & Rats, for Use with Management System IMS-310R, Pack of 5 Pumps
SDP
Catalog No. NC1746152
Encompass
iPRECIO SMP-310R pumps are advanced infusion devices for drug delivery in mice, rats and larger animals. With versatile programmable functions and a refillable reservoir, these implantable infusion pumps enable full control over dosing studies, including complex dosing protocols such as multiple bolus dosing, variable/constant delivery, escalating dose administration, and more. Access the pump reservoir in situ using the percutaneously accessible septum, which allows one to change solutions or refill the pump during the study. EtO sterilized.Set the infusion profile with the user-friendly iPRECIO management software and then download it to the pump’s memory chip. [Please note that the management software (IMS-310R) is sold separately.]iPRECIO SMP-310R:•For mice and larger animals•Wireless data communication•Refillable while in vivo•Reservoir volume: 130 µl•Flow rates: 0-10 µl/hr[For drug delivery in rats and larger animals, see also the iPRECIO SMP-200.]
Catalog No. NC1746152 Supplier Alzet Supplier No. 0006145
iPRECIO SMP-310R pumps are advanced infusion devices for drug delivery in mice, rats and larger animals. With versatile programmable functions and a refillable reservoir, these implantable infusion pumps enable full control over dosing studies, including complex dosing protocols such as multiple bolus dosing, variable/constant delivery, escalating dose administration, and more. Access the pump reservoir in situ using the percutaneously accessible septum, which allows one to change solutions or refill the pump during the study. EtO sterilized.Set the infusion profile with the user-friendly iPRECIO management software and then download it to the pump’s memory chip. [Please note that the management software (IMS-310R) is sold separately.]iPRECIO SMP-310R:•For mice and larger animals•Wireless data communication•Refillable while in vivo•Reservoir volume: 130 µl•Flow rates: 0-10 µl/hr[For drug delivery in rats and larger animals, see also the iPRECIO SMP-200.]