Learn More
Bioworld SMPB, 100 mg
Supplier:  Bioworld 218100171
Encompass_Preferred
SMPB, Succinimidyl-4-(p-maleimidophenyl) ButyratePrimary amine and sulfhydryl reactiveEnzyme-antibody conjugationSpacer armReferences: 1.) Kitagawa, T. and Aikawa, T. (1976) J. Biochem., 79, 223. 2.) Teale, J.M. and Kearney, J.F. (1986) J. Mol. Cell. Immunol. 2, 283-292.M.W. = 356.3
Catalog No. 50-199-265