Learn More
Aviva Systems Biology Corp SMPD3 Antibody - middle region (ARP49331_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49331_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMPD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SMPD3. Alternate names: FLJ22593, MGC138443, NSMASE2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60064