Learn More
ABclonal Technology SMPD4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A154731000UL
Encompass_Preferred
The protein encoded by this gene is a sphingomyelinase that catalyzes the hydrolysis of membrane sphingomyelin to form phosphorylcholine and ceramide. This gene is activated by DNA damage, cellular stress, and tumor necrosis factor, but it is downregulated by wild-type p53. The encoded protein localizes to the endoplasmic reticulum and Golgi network.
Catalog No. 50-001-06275